Merck
  • Polydopamine-reinforced magnetization of zeolitic imidazolate framework ZIF-7 for magnetic solid-phase extraction of polycyclic aromatic hydrocarbons from the air-water environment.
Polydopamine-reinforced magnetization of zeolitic imidazolate framework ZIF-7 for magnetic solid-phase extraction of polycyclic aromatic hydrocarbons from the air-water environment.
Journal of chromatography. A (2016-05-29)
Suling Zhang, Weixuan Yao, Jianbo Ying, Hongting Zhao
ABSTRACT
Zeolitic imidazolate frameworks (ZIFs) consist of metal nodes connected to imidazolate linkers, having both the properties of metal-organic frameworks (MOFs) and inorganic zeolites, such as controllable pore sizes, high porosity and surface areas, as well as exceptional thermal and chemical stability, thereby making them a class of attractive materials for diverse analytical applications. In this study, we reported a facile magnetization process of ZIF-7 (zinc benzimidazolate) for simultaneous magnetic extraction of polycyclic aromatic hydrocarbons (PAHs) by simply mixing ZIF-7 and polydopamine (PDA)-coated Fe3O4 nanoparticles (PDA@Fe3O4) in solutions. Functional groups (-OH and -NH2), provided by PDA as a highly efficient molecular linker, could attract and anchor ZIF-7 through noncovalent adsorption and covalent cross-link interactions, thereby promoting the complete magnetization of ZIFs and enhancing their stability and reusability. The bridging ligand benzimidazolate, could be bonded with PAHs because of its high surface area, large pores, accessible coordinative unsaturated sites (π-complexation), and π-π stacking action. This ZIF-based magnetic solid-phase extraction (SPE), coupled with gas chromatography/tandem mass spectrometry (GC/MS), was further evaluated for analysis of PAHs from rainwater and air samples of particulate matter less than 2.5μm in diameter (PM2.5). The main effective parameters, including ionic strength, solution pH, extraction time, desorption solvent and desorption time, were investigated, respectively. Under optimized conditions, the developed method based on Fe3O4@PDA/ZIF-7 gave detection limits of 0.71-5.79ng/L, and quantification limits of 2.50-19.2ng/L for PAHs, respectively. The relative standard deviations for intra-day and inter-day analyses were in the range of 3.1-9.1% and 6.1-12.7%, respectively. The PAHs founded in PM2.5 were in the range of 0.40-6.79ng/m(3). Good recoveries (>82%) with low relative standard deviations (≤9.2%) were achieved. This method was demonstrated to be an accurate, convenient and sensitive pretreatment procedure for trace analysis of environmental water and air samples.
Product Description
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, size 24 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 2, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 2, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assortment kit 4, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, needle size 23 ga, StableFlex fiber, for use with autosampler
Supelco
SPME StableFlex fiber assortment kit, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assortment kit 1, for use with autosampler, needle size 23 ga
Supelco
SPME StableFlex fiber assortment kit, for use with manual holder
Supelco
SPME fiber assortment kit 4, for use with manual holder, size 24 ga
Supelco
SPME fiber assortment kit 1, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assortment kit 5, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, metal alloy fiber, for use with manual holder
Supelco
SPME fiber assembly, Carbowax-Polyethylene Glycol (PEG) Coating, needle size 23 ga, df 60 μm(PEG, for use with autosampler
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with autosampler, needle size 24 ga
Supelco
SPME Portable Field Sampler, coating PDMS/DVB
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with manual holder
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 7 μm(PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly polyacrylate (PA), df 85 μm(PA, for use with manual holder, needle size 24 ga
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, for use with autosampler, needle size 23 ga
Supelco
SPME Sampling Stand, for use with 4 mL vials
Supelco
SPME Fiber Assembly Polydimethylsiloxane/Divinylbenzene (PDMS/DVB), df 65 μm(PDMS/DVB, needle size 24 ga, StableFlex, for use with autosampler