Merck
  • Inhibition of macrophage Ca(2+)-independent phospholipase A2 by bromoenol lactone and trifluoromethyl ketones.
Inhibition of macrophage Ca(2+)-independent phospholipase A2 by bromoenol lactone and trifluoromethyl ketones.
The Journal of biological chemistry (1995-01-06)
E J Ackermann, K Conde-Frieboes, E A Dennis
ABSTRACT
A novel Ca(2+)-independent phospholipase A2 (PLA2) has recently been purified from the murine macrophage-like cell line P388D1 (Ackermann, E. J., Kempner, E. S., and Dennis, E. A. (1994) J. Biol. Chem. 269, 9227-9233). This enzyme is now shown to be inhibited by palmitoyl trifluoromethyl ketone (PACOCF3), arachidonyl trifluoromethyl ketone (AACOCF3), and a bromoenol lactone (BEL). Both PACOCF3 and AACOCF3 were found to inhibit the macrophage PLA2 in a concentration-dependent manner. PACOCF3 was found to be approximately 4-fold more potent than AACOCF3, with IC50 values of 3.8 microM (0.0075 mol fraction) and 15 microM (0.028 mol fraction), respectively. Reaction progress curves in the presence of either inhibitor were found to be linear, and the PACOCF3.PLA2 complex rapidly dissociated upon dilution. BEL was also found to inhibit the macrophage PLA2 in a concentration-dependent manner, with half-maximal inhibition observed at 60 nM after a 5-min preincubation at 40 degrees C. Inhibition was not reversed after extensive dilution of the enzyme into assay buffer. Treatment of the PLA2 with BEL resulted in a linear, time-dependent inactivation of activity, and the rate of this inactivation was diminished in the presence of PACOCF3. In addition, PLA2 treated with [3H]BEL resulted in the covalent labeling of a major band at M(r) 80,000. Inactivation of the PLA2 by 5,5'-dithiobis(2-nitrobenzoic acid) prior to treatment with [3H]BEL resulted in the near complete lack of labeling consistent with covalent irreversible suicide inhibition of the enzyme. The labeling of a M(r) 80,000 band rather than a M(r) 40,000 band upon treatment with [3H]BEL distinguishes the macrophage Ca(2+)-independent PLA2 from a previously identified myocardial Ca(2+)-independent PLA2 and provides strong evidence that the M(r) 80,000 protein is the catalytic subunit.
Sigma-Aldrich
Bromoenol lactone, ≥98% (TLC)
Sigma-Aldrich
Arachidonyl trifluoromethyl ketone, oil, ≥97% (NMR)