Merck
  • Photocrosslinking of Polyglycidol and Its Derivative: Route to Thermoresponsive Hydrogels.
Photocrosslinking of Polyglycidol and Its Derivative: Route to Thermoresponsive Hydrogels.
Photochemistry and photobiology (2017-08-03)
Alicja Utrata-Wesołek, Iwona Żymełka-Miara, Agnieszka Kowalczuk, Barbara Trzebicka, Andrzej Dworak
ABSTRACT
Hydrogels of biologically well-tolerated, high-molar-mass polyglycidol (PGl) and its thermoresponsive derivative poly(glycidol-co-ethyl glycidyl carbamate) have been obtained by direct UV crosslinking in the solid state. Polymers with molar masses up to 1.45 × 10
Sigma-Aldrich
Dibutyltin dilaurate, 95%
Sigma-Aldrich
Ethyl isocyanate, 98%