Merck
  • Decanoic acid suppresses proliferation and invasiveness of human trophoblast cells by disrupting mitochondrial function.
Decanoic acid suppresses proliferation and invasiveness of human trophoblast cells by disrupting mitochondrial function.
Toxicology and applied pharmacology (2017-12-19)
Changwon Yang, Whasun Lim, Fuller W Bazer, Gwonhwa Song
ABSTRACT
Decanoic acid (DA) is a medium-chain fatty acid used in the manufacture of various products including plastics, cosmetics, and lubricants. In addition to antiviral and antibacterial effects, DA's, reported biological activities include regulation of signaling pathways and redox homeostasis in various human cell types. The influence of DA on functional properties of human trophoblasts, including proliferation, invasion and apoptosis is currently unknown. In the present study, we evaluated the anti-proliferative and anti-invasive effects of DA on the human trophoblast cell line HTR8/SVneo. In addition, DA induced oxidative stress, as evidenced by generation of reactive oxygen species (ROS) and induction of lipid peroxidation (LPO). This oxidative stress was accompanied by activation of the mitochondria-dependent apoptotic pathway in HTR8/SVneo cells. We also observed elevated mitochondrial Ca2+, and loss of mitochondrial membrane potential in response to DA treatment. Chelation of mitochondrial Ca2+ using BAPTA-AM rescued cellular proliferation suppressed by DA. We also verified that signaling proteins including AKT, P70S6K, S6, and ERK1/2 and their targets were significantly reduced in HTR8/SVneo cells by DA treatment. Pre-treatment of cells with selective inhibitors of AKT (LY294002) and ERK1/2 (U0126) revealed that the AKT and ERK1/2 signaling pathways regulated by DA displayed cross-talk in HTR8/SVneo cells. Collectively, these results suggest that personal products containing DA will have harmful effects on human trophoblasts, and could cause implantation and placentation failure during early pregnancy.
Sigma-Aldrich
Mitochondria Staining Kit, 1 kit sufficient for 40 tests (of 5 mL cell suspensions), 1 kit sufficient for 200 tests (of 1 mL cell suspensions)
Pricing and availability is not currently available.
Supelco
Decanoic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Pricing and availability is not currently available.