Merck
Na
Frontiers in molecular neuroscience (2018-02-08)
Corinna Martin, Carolin Stoffer, Milad Mohammadi, Julian Hugo, Enrico Leipold, Beatrice Oehler, Heike L Rittner, Robert Blum
ABSTRACT
Oxidized phospholipids (OxPL) like oxidized 1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphocholine (OxPAPC) were recently identified as novel proalgesic targets in acute and chronic inflammatory pain. These endogenous chemical irritants are generated in inflamed tissue and mediate their pain-inducing function by activating the transient receptor potential channels TRPA1 and TRPV1 expressed in sensory neurons. Notably, prototypical therapeutics interfering with OxPL were shown to inhibit TRP channel activation and pain behavior. Here, we asked how OxPL excite primary sensory neurons of dorsal root ganglia (DRG neurons from mice of either sex). Acute stimulation of sensory neurons with the prototypical OxPL 1-palmitoyl-2-glutaryl-
Sigma-Aldrich
BCTC, ≥98% (HPLC)