Merck
SK4 K
EMBO molecular medicine (2017-02-22)
Shiraz Haron-Khun, David Weisbrod, Hanna Bueno, Dor Yadin, Joachim Behar, Asher Peretz, Ofer Binah, Edith Hochhauser, Michael Eldar, Yael Yaniv, Michael Arad, Bernard Attali
ABSTRACT
Catecholaminergic polymorphic ventricular tachycardia (CPVT) is a stress-provoked ventricular arrhythmia, which also manifests sinoatrial node (SAN) dysfunction. We recently showed that SK4 calcium-activated potassium channels are important for automaticity of cardiomyocytes derived from human embryonic stem cells. Here SK4 channels were identified in human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CMs) from healthy and CPVT2 patients bearing a mutation in calsequestrin 2 (CASQ2-D307H) and in SAN cells from WT and CASQ2-D307H knock-in (KI) mice. TRAM-34, a selective blocker of SK4 channels, prominently reduced delayed afterdepolarizations and arrhythmic Ca
Sigma-Aldrich
Anti-KCNN4 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-KCNN4 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution