Merck
  • Palladium(II) acetate in pyridine as an effective catalyst for highly regioselective hydroselenation of alkynes.
Palladium(II) acetate in pyridine as an effective catalyst for highly regioselective hydroselenation of alkynes.
The Journal of organic chemistry (2005-01-18)
Ikuyo Kamiya, Etsuyo Nishinaka, Akiya Ogawa
ABSTRACT
A highly regioselective hydroselenation of terminal alkynes with benzeneselenol can be achieved by the combination of palladium acetate and pyridine, providing the corresponding terminal alkenes, (i.e., 2-phenylseleno-1-alkenes) as a sole product. In this hydroselenation, pyridine may act as a suitable ligand for active palladium intermediates.
Sigma-Aldrich
Palladium(II) acetate, ≥99.9% trace metals basis
Sigma-Aldrich
Palladium(II) acetate, reagent grade, 98%
Sigma-Aldrich
Palladium(II) Acetate ChemBeads