Merck
  • HoxC5 and miR-615-3p target newly evolved genomic regions to repress hTERT and inhibit tumorigenesis.
HoxC5 and miR-615-3p target newly evolved genomic regions to repress hTERT and inhibit tumorigenesis.
Nature communications (2018-01-10)
TingDong Yan, Wen Fong Ooi, Aditi Qamra, Alice Cheung, DongLiang Ma, Gopinath Meenakshi Sundaram, Chang Xu, Manjie Xing, LaiFong Poon, Jing Wang, Yan Ping Loh, Jess Hui Jie Ho, Joscelyn Jun Quan Ng, Muhammad Khairul Ramlee, Luay Aswad, Steve G Rozen, Sujoy Ghosh, Frederic A Bard, Prabha Sampath, Vinay Tergaonkar, James O J Davies, Jim R Hughes, Eyleen Goh, Xuezhi Bi, Melissa Jane Fullwood, Patrick Tan, Shang Li
ABSTRACT
The repression of telomerase activity during cellular differentiation promotes replicative aging and functions as a physiological barrier for tumorigenesis in long-lived mammals, including humans. However, the underlying mechanisms remain largely unclear. Here we describe how miR-615-3p represses hTERT expression. mir-615-3p is located in an intron of the HOXC5 gene, a member of the highly conserved homeobox family of transcription factors controlling embryogenesis and development. Unexpectedly, we found that HoxC5 also represses hTERT expression by disrupting the long-range interaction between hTERT promoter and its distal enhancer. The 3'UTR of hTERT and its upstream enhancer region are well conserved in long-lived primates. Both mir-615-3p and HOXC5 are activated upon differentiation, which constitute a feed-forward loop that coordinates transcriptional and post-transcriptional repression of hTERT during cellular differentiation. Deregulation of HOXC5 and mir-615-3p expression may contribute to the activation of hTERT in human cancers.
Sigma-Aldrich
MC1568, ≥97% (HPLC)
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Anti-HOXC5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-PBX4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-RNA polymerase II Antibody, clone CTD4H8, clone CTD4H8, Upstate®, from mouse
Sigma-Aldrich
Trichostatin A, Ready Made Solution, 5 mM in DMSO, from Streptomyces sp.