Merck
  • Migrating Platelets Are Mechano-scavengers that Collect and Bundle Bacteria.
Migrating Platelets Are Mechano-scavengers that Collect and Bundle Bacteria.
Cell (2017-12-02)
Florian Gaertner, Zerkah Ahmad, Gerhild Rosenberger, Shuxia Fan, Leo Nicolai, Benjamin Busch, Gökce Yavuz, Manja Luckner, Hellen Ishikawa-Ankerhold, Roman Hennel, Alexandre Benechet, Michael Lorenz, Sue Chandraratne, Irene Schubert, Sebastian Helmer, Bianca Striednig, Konstantin Stark, Marek Janko, Ralph T Böttcher, Admar Verschoor, Catherine Leon, Christian Gachet, Thomas Gudermann, Michael Mederos Y Schnitzler, Zachary Pincus, Matteo Iannacone, Rainer Haas, Gerhard Wanner, Kirsten Lauber, Michael Sixt, Steffen Massberg
ABSTRACT
Blood platelets are critical for hemostasis and thrombosis and play diverse roles during immune responses. Despite these versatile tasks in mammalian biology, their skills on a cellular level are deemed limited, mainly consisting in rolling, adhesion, and aggregate formation. Here, we identify an unappreciated asset of platelets and show that adherent platelets use adhesion receptors to mechanically probe the adhesive substrate in their local microenvironment. When actomyosin-dependent traction forces overcome substrate resistance, platelets migrate and pile up the adhesive substrate together with any bound particulate material. They use this ability to act as cellular scavengers, scanning the vascular surface for potential invaders and collecting deposited bacteria. Microbe collection by migrating platelets boosts the activity of professional phagocytes, exacerbating inflammatory tissue injury in sepsis. This assigns platelets a central role in innate immune responses and identifies them as potential targets to dampen inflammatory tissue damage in clinical scenarios of severe systemic infection.
Sigma-Aldrich
Picropodophyllotoxin, ≥96% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Integrin αVβ3 Antibody, clone LM609, clone LM609, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Hexamethyldisilazane, Wacker Chemie AG, ≥97.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.