Merck
Regulation of Arthritis Severity by the Acid Sphingomyelinase.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2017-10-17)
Nadine Beckmann, Katrin Anne Becker, Silke Walter, Jan U Becker, Melanie Kramer, Gabriele Hessler, Stefanie Weber, Joachim R Göthert, Klaus Fassbender, Erich Gulbins, Alexander Carpinteiro
ABSTRACT
Rheumatoid arthritis is a chronic autoimmune disease hallmarked by inflammation in synovial joints. Treatment is hampered by the lack of a cure and current disease-modifying drugs are associated with potentially severe toxicities. We investigated arthritis severity by measuring joint swelling and pro-inflammatory cytokine production in a murine experimental model of inflammatory arthritis (antigen-induced arthritis). We analyzed acid sphingomyelinase knock-out mice and wild-type littermates, as well as mice treated with the pharmacological acid sphingomyelinase inhibitor amitriptyline. Genetic ablation or pharmacological inhibition of acid sphingomyelinase reduced joint swelling and levels of pro-inflammatory cytokines in the arthritic joint. We identified acid sphingomyelinase as a novel druggable target in rheumatoid arthritis. Functional inhibitors of acid sphingomyelinase have been clinically used for decades, are well tolerated and suitable for long-term treatment. They would be immediately available for clinical development as a novel rheumatoid arthritis therapy.
Sigma-Aldrich
Amitriptyline hydrochloride, ≥98% (TLC), powder