Merck
  • Integrin α4β1 and TLR4 Cooperate to Induce Fibrotic Gene Expression in Response to Fibronectin's EDA Domain.
Integrin α4β1 and TLR4 Cooperate to Induce Fibrotic Gene Expression in Response to Fibronectin's EDA Domain.
The Journal of investigative dermatology (2017-08-27)
Rhiannon M Kelsh-Lasher, Anthony Ambesi, Ceyda Bertram, Paula J McKeown-Longo
ABSTRACT
Alternative splicing of fibronectin increases expression of the EDA+ isoform of fibronectin (EDA+Fn), a damage-associated molecular pattern molecule, which promotes fibro-inflammatory disease through the activation of toll-like receptors. Our studies indicate that the fibronectin EDA domain drives two waves of gene expression in human dermal fibroblasts. The first wave, seen at 2 hours, consisted of inflammatory genes, VCAM1, and tumor necrosis factor. The second wave, evaluated at 24 hours, was composed of the fibrosis-associated cytokines IL-10 and IL-13 and extracellular matrix genes fibronectin and osteopontin. Gene expression was coordinately regulated by the α4β1 integrin and the innate immune receptor toll-like receptor 4. Additionally, we found a significant toll-like receptor 4/α4β1-dependent enrichment in the ratio of EDA+Fn to total fibronectin in response to EDA, consistent with EDA+Fn initiating further production of EDA+Fn. Our data also suggest that the EDA/α4β1 integrin interaction primes the cell for an enhanced response to toll-like receptor 4 ligands. Our studies provide evidence that remodeling of the fibronectin matrix in injured or diseased tissue elicits an EDA-dependent fibro-inflammatory response in dermal fibroblasts. The data suggest a paradigm of damage-associated molecular pattern-based signaling whereby damage-associated molecular pattern binding integrins cooperate with innate immune receptors to stimulate inflammation and fibrosis.
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Anti-Integrin α4 Antibody, clone P1H4, clone P1H4, Chemicon®, from mouse