Merck
  • Peroxisome proliferator-activated receptor gamma (PPARγ) regulates lactase expression and activity in the gut.
Peroxisome proliferator-activated receptor gamma (PPARγ) regulates lactase expression and activity in the gut.
EMBO molecular medicine (2017-09-28)
Mathurin Fumery, Silvia Speca, Audrey Langlois, Anne-Marie Davila, Caroline Dubuquoy, Marta Grauso, Anthony Martin Mena, Martin Figeac, Daniel Metzger, Christel Rousseaux, Jean-Frederic Colombel, Laurent Dubuquoy, Pierre Desreumaux, Benjamin Bertin
ABSTRACT
Lactase (LCT) deficiency affects approximately 75% of the world's adult population and may lead to lactose malabsorption and intolerance. Currently, the regulation of LCT gene expression remains poorly known. Peroxisome proliferator activator receptorγ (PPARγ) is a key player in carbohydrate metabolism. While the intestine is essential for carbohydrate digestion and absorption, the role of PPARγ in enterocyte metabolic functions has been poorly investigated. This study aims at characterizing PPARγ target genes involved in intestinal metabolic functions. In microarray analysis, the LCT gene was the most upregulated by PPARγ agonists in Caco-2 cells. We confirmed that PPARγ agonists were able to increase the expression and activity of LCT both
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Linoleic acid, conjugated
Sigma-Aldrich
Pioglitazone hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Pioglitazone