- Changes in Gene Expression in Space and Time Orchestrate Environmentally Mediated Shaping of Root Architecture.
Changes in Gene Expression in Space and Time Orchestrate Environmentally Mediated Shaping of Root Architecture.
The Plant cell (2017-09-13)
Liam Walker, Clare Boddington, Dafyd Jenkins, Ying Wang, Jesper T Grønlund, Jo Hulsmans, Sanjeev Kumar, Dhaval Patel, Jonathan D Moore, Anthony Carter, Siva Samavedam, Giovanni Bonomo, David S Hersh, Gloria M Coruzzi, Nigel J Burroughs, Miriam L Gifford
PMID28893852
ABSTRACT
Shaping of root architecture is a quintessential developmental response that involves the concerted action of many different cell types, is highly dynamic, and underpins root plasticity. To determine to what extent the environmental regulation of lateral root development is a product of cell-type preferential activities, we tracked transcriptomic responses to two different treatments that both change root development in
Sigma-Aldrich
Murashige and Skoog Basal Salt Micronutrient Solution, 10 ×, liquid, suitable for plant cell culture