Merck
  • Mitochondrial Fission Promotes the Continued Clearance of Apoptotic Cells by Macrophages.
Mitochondrial Fission Promotes the Continued Clearance of Apoptotic Cells by Macrophages.
Cell (2017-09-26)
Ying Wang, Manikandan Subramanian, Arif Yurdagul, Valéria C Barbosa-Lorenzi, Bishuang Cai, Jaime de Juan-Sanz, Timothy A Ryan, Masatoshi Nomura, Frederick R Maxfield, Ira Tabas
ABSTRACT
Clearance of apoptotic cells (ACs) by phagocytes (efferocytosis) prevents post-apoptotic necrosis and dampens inflammation. Defective efferocytosis drives important diseases, including atherosclerosis. For efficient efferocytosis, phagocytes must be able to internalize multiple ACs. We show here that uptake of multiple ACs by macrophages requires dynamin-related protein 1 (Drp1)-mediated mitochondrial fission, which is triggered by AC uptake. When mitochondrial fission is disabled, AC-induced increase in cytosolic calcium is blunted owing to mitochondrial calcium sequestration, and calcium-dependent phagosome formation around secondarily encountered ACs is impaired. These defects can be corrected by silencing the mitochondrial calcium uniporter (MCU). Mice lacking myeloid Drp1 showed defective efferocytosis and its pathologic consequences in the thymus after dexamethasone treatment and in advanced atherosclerotic lesions in fat-fed Ldlr-/- mice. Thus, mitochondrial fission in response to AC uptake is a critical process that enables macrophages to clear multiple ACs and to avoid the pathologic consequences of defective efferocytosis in vivo.
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), lyophilized powder
Sigma-Aldrich
Diluent C for General Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)
USP
Collagenase I, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Neomycin solution, with 10 mg/mL neomycin in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Roche
In Situ Cell Death Detection Kit, TMR red, sufficient for ≤50 tests