Merck
  • Chronic antidepressant potentiates spontaneous activity of dorsal raphe serotonergic neurons by decreasing GABA
Chronic antidepressant potentiates spontaneous activity of dorsal raphe serotonergic neurons by decreasing GABA
Scientific reports (2017-10-21)
Nozomi Asaoka, Naoya Nishitani, Haruko Kinoshita, Hiroyuki Kawai, Norihiro Shibui, Kazuki Nagayasu, Hisashi Shirakawa, Takayuki Nakagawa, Shuji Kaneko
ABSTRACT
Spontaneous activity of serotonergic neurons of the dorsal raphe nucleus (DRN) regulates mood and motivational state. Potentiation of serotonergic function is one of the therapeutic strategies for treatment of various psychiatric disorders, such as major depression, panic disorder and obsessive-compulsive disorder. However, the control mechanisms of the serotonergic firing activity are still unknown. In this study, we examined the control mechanisms for serotonergic spontaneous activity and effects of chronic antidepressant administration on these mechanisms by using modified ex vivo electrophysiological recording methods. Serotonergic neurons remained firing even in the absence of glutamatergic and GABAergic ionotropic inputs, while blockade of L-type voltage dependent Ca
Sigma-Aldrich
DNQX, ≥98% (TLC)
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid