Merck
  • High Yield of Adult Oligodendrocyte Lineage Cells Obtained from Meningeal Biopsy.
High Yield of Adult Oligodendrocyte Lineage Cells Obtained from Meningeal Biopsy.
Frontiers in pharmacology (2017-10-28)
Sissi Dolci, Annachiara Pino, Valeria Berton, Pau Gonzalez, Alice Braga, Marta Fumagalli, Elisabetta Bonfanti, Giorgio Malpeli, Francesca Pari, Stefania Zorzin, Clelia Amoroso, Denny Moscon, Francisco J Rodriguez, Guido Fumagalli, Francesco Bifari, Ilaria Decimo
ABSTRACT
Oligodendrocyte loss can lead to cognitive and motor deficits. Current remyelinating therapeutic strategies imply either modulation of endogenous oligodendrocyte precursors or transplantation of in vitro expanded oligodendrocytes. Cell therapy, however, still lacks identification of an adequate source of oligodendrocyte present in adulthood and able to efficiently produce transplantable cells. Recently, a neural stem cell-like population has been identified in meninges. We developed a protocol to obtain high yield of oligodendrocyte lineage cells from one single biopsy of adult rat meningeal tissue. From 1 cm2 of adult rat spinal cord meninges, we efficiently expanded a homogenous culture of 10 millions of meningeal-derived oligodendrocyte lineage cells in a short period of time (approximately 4 weeks). Meningeal-derived oligodendrocyte lineage cells show typical mature oligodendrocyte morphology and express specific oligodendrocyte markers, such as galactosylceramidase and myelin basic protein. Moreover, when transplanted in a chemically demyelinated spinal cord model, meningeal-derived oligodendrocyte lineage cells display in vivo-remyelinating potential. This oligodendrocyte lineage cell population derives from an accessible and adult source, being therefore a promising candidate for autologous cell therapy of demyelinating diseases. In addition, the described method to differentiate meningeal-derived neural stem cells into oligodendrocyte lineage cells may represent a valid in vitro model to dissect oligodendrocyte differentiation and to screen for drugs capable to promote oligodendrocyte regeneration.
Sigma-Aldrich
Monoclonal Anti-Oligodendrocyte Marker O4 antibody produced in mouse, clone O4, purified immunoglobulin, lyophilized powder
Sigma-Aldrich
Monoclonal Anti-Neurofilament 160 antibody produced in mouse, clone NN18, ascites fluid
Sigma-Aldrich
Monoclonal Anti-MBP antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2829, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-O4 Antibody, clone 81, clone 81 (mAB O4), Chemicon®, from mouse