Merck
  • Liquid chromatography-tandem mass spectrometry multiresidue method for the analysis of quaternary ammonium compounds in cheese and milk products: Development and validation using the total error approach.
Liquid chromatography-tandem mass spectrometry multiresidue method for the analysis of quaternary ammonium compounds in cheese and milk products: Development and validation using the total error approach.
Journal of chromatography. A (2017-08-31)
Kahina Slimani, Aurélie Féret, Yvette Pirotais, Pierre Maris, Jean-Pierre Abjean, Dominique Hurtaud-Pessel
ABSTRACT
Quaternary ammonium compounds (QACs) are both cationic surfactants and biocidal substances widely used as disinfectants in the food industry. A sensitive and reliable method for the analysis of benzalkonium chlorides (BACs) and dialkyldimethylammonium chlorides (DDACs) has been developed that enables the simultaneous quantitative determination of ten quaternary ammonium residues in dairy products below the provisional maximum residue level (MRL), set at 0.1mgkg
Sigma-Aldrich
Didecyldimethylammonium bromide, 98%
Sigma-Aldrich
Didodecyldimethylammonium bromide, 98%
Sigma-Aldrich
Dimethylditetradecylammonium bromide, ≥97.0% (NT)
Sigma-Aldrich
Dihexadecyldimethylammonium bromide, 97%
Sigma-Aldrich
Trifluoroacetic anhydride, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzyldimethyldodecylammonium chloride, ≥99.0% (AT)
Sigma-Aldrich
Benzyldimethyloctylammonium chloride, ≥96.0% (AT)
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)