Merck
  • Reduction of body-weight gain enhances in vitro embryo production in overfed superovulated dairy heifers.
Reduction of body-weight gain enhances in vitro embryo production in overfed superovulated dairy heifers.
Reproduction (Cambridge, England) (2006-04-06)
S Freret, B Grimard, A A Ponter, C Joly, C Ponsart, P Humblot
ABSTRACT
The aim of our study was to test whether a reduction in dietary intake could improve in vitro embryo production in superovulated overfed dairy heifers. Cumulus-oocyte complexes of 16 Prim' Holstein heifers (14 +/- 1 months old) were collected by ovum pick-up (OPU), every 2 weeks following superovulation treatment with 250 microg FSH, before being matured and fertilized in vitro. Embryos were cultured in Synthetic Oviduct Fluid medium for 7 days. Heifers were fed with hay, soybean meal, barley, minerals and vitamins. From OPU 1 to 4 (period 1), all heifers received individually for 8 weeks a diet formulated for a 1000 g/day live-weight gain. From OPU 5 to 8 (period 2), the heifers were allocated to one of two diets (1000 or 600 g/day) for 8 weeks. Heifers' growth rates were monitored and plasma concentrations of metabolites, metabolic and reproductive hormones were measured each week. Mean live-weight gain observed during period 1 was 950 +/- 80 g/day (n = 16). In period 2 it was 730 +/- 70 (n = 8) and 1300 +/- 70 g/day (n = 8) for restricted and overfed groups respectively. When comparing period 1 and period 2 within groups, significant differences were found. In the restricted group, a higher blastocyst rate, greater proportions of grade 1-3 and grade 1 embryos, associated with higher estradiol at OPU and lower glucose and beta-hydroxybutyrate, were observed in period 2 compared with period 1. Moreover, after 6 weeks of dietary restriction (OPU 7), numbers of day 7 total embryos, blastocysts and grade 1-3 embryos had significantly increased. On the contrary, in the overfed group, we observed more <8 mm follicles 2 days before superovulation treatment, higher insulin and IGF-I and lower nonesterified fatty acids in period 2 compared with period 1 (no significant difference between periods for embryo production). After 6 weeks of 1300 g/day live-weight gain (OPU 7), embryo production began to decrease. Whatever the group, oocyte collection did not differ between period 1 and 2. These data suggest that following a period of overfeeding, a short-term dietary intake restriction (6 weeks in our study) may improve blastocyst production and embryo quality when they are low. However, nutritional recommendations aiming to optimize both follicular growth and embryonic development may be different.
Sigma-Aldrich
Gentamicin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
(−)-Epinephrine
Sigma-Aldrich
Mineral oil, suitable for preparation of Nujol mulls for infrared spectroscopy, light oil
Sigma-Aldrich
β-Estradiol, powder, γ-irradiated, suitable for cell culture