- Increased 15-PGDH expression leads to dysregulated resolution responses in stromal cells from patients with chronic tendinopathy.
Increased 15-PGDH expression leads to dysregulated resolution responses in stromal cells from patients with chronic tendinopathy.
Scientific reports (2017-09-10)
Stephanie G Dakin, Lucy Ly, Romain A Colas, Udo Oppermann, Kim Wheway, Bridget Watkins, Jesmond Dalli, Andrew J Carr
PMID28887458
ABSTRACT
The mechanisms underpinning the failure of inflammation to resolve in diseased musculoskeletal soft tissues are unknown. Herein, we studied bioactive lipid mediator (LM) profiles of tendon-derived stromal cells isolated from healthy donors and patients with chronic tendinopathy. Interleukin(IL)-1β treatment markedly induced prostaglandin biosynthesis in diseased compared to healthy tendon cells, and up regulated the formation of several pro-resolving mediators including 15-epi-LXA