Merck
  • A novel highly selective ligand for separation of actinides and lanthanides in the nuclear fuel cycle. Experimental verification of the theoretical prediction.
A novel highly selective ligand for separation of actinides and lanthanides in the nuclear fuel cycle. Experimental verification of the theoretical prediction.
Dalton transactions (Cambridge, England : 2003) (2017-08-03)
H V Lavrov, N A Ustynyuk, P I Matveev, I P Gloriozov, S S Zhokhov, M Yu Alyapyshev, L I Tkachenko, I G Voronaev, V A Babain, S N Kalmykov, Yu A Ustynyuk
ABSTRACT
We have predicted earlier by DFT simulation that tridentate O,N,O-donor cyclic dilactams (B) belonging to the family of pyridine-2,6-dicarboxamides are much more selective and efficient extractants for the separation of lanthanides and actinides than open-structure pyridine-2,6-dicarboxamides due to the higher degree of "ligand preorganization". In the present work, three new ligands of type (B) were synthesized. Extraction experiments showed that, in line with the data from DFT simulation, these ligands have 5-6-fold higher selectivity for the separation of an Am
Sigma-Aldrich
3-Nitrobenzotrifluoride, 97%