Merck
  • Role of canonical transient receptor potential channel-3 in acetylcholine-induced mouse airway smooth muscle cell proliferation.
Role of canonical transient receptor potential channel-3 in acetylcholine-induced mouse airway smooth muscle cell proliferation.
Life sciences (2017-08-15)
Xiao-Xu Chen, Jia-Hua Zhang, Bin-Hua Pan, Hui-Li Ren, Xiu-Ling Feng, Jia-Ling Wang, Jun-Hua Xiao
ABSTRACT
Canonical transient receptor potential channel-3 (TRPC3)-encoded Ca Primary mouse ASMCs were cultured with or without ACh treatment, then cell viability, TRPC3 expression, NSCC currents and [Ca TRPC3 blocker Gd Our data suggested ACh could induce ASMC proliferation, and TRPC3 may be involved in ACh-induced ASMC proliferation that occurs with airway remodeling.
Sigma-Aldrich
Acetylcholine bromide, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting human TRPC3