Merck
Aspartic Proteases and Major Cell Wall Components in
Frontiers in cellular and infection microbiology (2017-10-07)
Marcin Zawrotniak, Oliwia Bochenska, Justyna Karkowska-Kuleta, Karolina Seweryn-Ozog, Wataru Aoki, Mitsuyoshi Ueda, Andrzej Kozik, Maria Rapala-Kozik
ABSTRACT
Neutrophils use different mechanisms to cope with pathogens that invade the host organism. The most intriguing of these responses is a release of neutrophil extracellular traps (NETs) composed of decondensed chromatin and granular proteins with antimicrobial activity. An important potential target of NETs is
Sigma-Aldrich
Piceatannol, powder
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%
Sigma-Aldrich
PP2, ≥98% (HPLC)