Merck
  • IN0523 (Urs-12-ene-3α,24β-diol) a plant based derivative of boswellic acid protect Cisplatin induced urogenital toxicity.
IN0523 (Urs-12-ene-3α,24β-diol) a plant based derivative of boswellic acid protect Cisplatin induced urogenital toxicity.
Toxicology and applied pharmacology (2017-01-26)
Amarinder Singh, S Arvinda, Surjeet Singh, Jyotsna Suri, Surinder Koul, Dilip M Mondhe, Gurdarshan Singh, Ram Vishwakarma
ABSTRACT
The limiting factor for the use of Cisplatin in the treatment of different type of cancers is its toxicity and more specifically urogenital toxicity. Oxidative stress is a well-known phenomenon associated with Cisplatin toxicity. However, in Cisplatin treated group, abnormal animal behavior, decreased body weight, cellular and sub-cellular changes in the kidney and sperm abnormality were observed. Our investigation revealed that Cisplatin when administered in combination with a natural product derivative (Urs-12-ene-3α,24β-diol, labeled as IN0523) resulted in significant restoration of body weight and protection against the pathological alteration caused by Cisplatin to kidney and testis. Sperm count and motility were significantly restored near to normal. Cisplatin caused depletion of defense system i.e. glutathione peroxidase, catalase and superoxide dismutase, which were restored close to normal by treatment of IN0523. Reduction in excessive lipid peroxidation induced by Cisplatin was also found by treatment with IN0523. The result suggests that IN0523 is a potential candidate for ameliorating Cisplatin induced toxicity in the kidney and testes at a dose of 100mg/kg p.o. via inhibiting the oxidative stress/redox status imbalance and may be improving the efflux mechanism.
Sigma-Aldrich
Cerium(IV) sulfate