Merck
  • The nature of the light absorption and emission transitions of 4-hydroxybenzophenone in different solvents. A combined computational and experimental study.
The nature of the light absorption and emission transitions of 4-hydroxybenzophenone in different solvents. A combined computational and experimental study.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2017-01-20)
Francesco Barsotti, Giovanni Ghigo, Silvia Berto, Davide Vione
ABSTRACT
The photophysics and photochemistry of 4-hydroxybenzophenone (4HOBP) are interesting because they can give some insight into the behavior of humic material. Here we show that 4HOBP has a number of fluorescence peaks: (i) an intense one at excitation/emission wavelengths Ex/Em ∼ 200-230/280-370 nm, likely due to an excitation transition from S
Sigma-Aldrich
4-Hydroxybenzophenone, 98%