Merck
  • Advanced glycation end products interfere with gastric smooth muscle contractile marker expression via the AGE/RAGE/NF-κB pathway.
Advanced glycation end products interfere with gastric smooth muscle contractile marker expression via the AGE/RAGE/NF-κB pathway.
Experimental and molecular pathology (2016-12-13)
Ting Yu, Yongping Zheng, Yun Wang, Wenjie Xiong, Lin Lin
ABSTRACT
Excessive production of advanced glycation end products (AGE) has been implicated in the pathogenesis of diabetic complications. Smooth muscle (SM) phenotype transition is involved in diabetes-associated gastric motility dysfunction. We investigated whether AGE interfere with gastric antral SM contractile marker expression. Sixteen Sprague-Dawley rats were randomly divided into control and streptozotocin-induced diabetic groups. Sixteen weeks after streptozotocin administration, gastric antral SM strip contractility in the groups were measured. The gastric tissue expression of AGE was tested. Primary cultured gastric smooth muscle cells (SMCs) were used in complementary in vitro studies. In the presence and absence of AGE, SMCs were transfected with myocardin plasmid or treated with nuclear factor-κB (NF-κB) inhibitor or anti-RAGE antibody. Diabetic rats showed weakness of SM strip contractility and decreased expression of SM contractile marker genes (myosin heavy chains [MHC], α-actin, calponin) as compared with the control group. Gastric antral SM layer Nε-(carboxymethyl) lysine (CML) level, the major AGE compound, were increased in the diabetic rats. AGE downregulated SM contractile markers and myocardin expression in a concentration-dependent manner. Myocardin overexpression prevented these results. AGE treatment activated NF-κB in SMCs. The NF-κB inhibitor BAY 11-7082 and anti-RAGE antibody blocked the effects of AGE on myocardin downregulation. AGE may induce the development of gastric dysmotility by downregulating SM contractile proteins and myocardin expression via the AGE/RAGE/NF-κB pathway.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O