Merck
  • Enhanced expression of lipase I from Galactomyces geotrichum by codon optimisation in Pichia pastoris.
Enhanced expression of lipase I from Galactomyces geotrichum by codon optimisation in Pichia pastoris.
Protein expression and purification (2017-06-07)
Hanzhen Qiao, Wenfei Zhang, Wutai Guan, Fang Chen, Shihai Zhang, Zixiao Deng
ABSTRACT
Relatively poor heterologous protein yields have limited the commerical applications of Galactomyces geotrichum lipase I (GGl I) efficacy trials. To address this, we have redesigned the GGl I gene to preferentially match codon frequencies of Pichia pastoris (P. pastoris) while retaining the same amino acid sequence. The wild type and codon optimised GGl I (GGl I-wt and GGl I-op) were synthesised and cloned into pPICZαA with an N-terminal 6 × His tag sequence and expressed in P. pastoris X 33. The hydrolytic activity of GGl I-op was 150 U/mL, whereas the activity of the GGl I-wt could not be detected. GGl I-op recombinant proteins were purified by Ni-affinity chromatography and then characterised. The identity and purity of GGl I were confirmed by SDS-PAGE, MALDI-TOF mass spectrometry and Western blot analysis. Enzymatic deglycosylation was used to show that the lipase is a glycosylated protein, containing ∼10% sugar. The molecular weight (MW) of the GGl I secreted by recombinant P. pastoris was approximated at 63 kDa. The optimum pH and temperature of the recombinant lipase were 8.0 and 35 °C, respectively. The enzyme was active over a broad pH range (7.0-9.0) and temperature range (20 °C-45 °C). The lipase showed high activity toward medium- and long-chain fatty acid methyl esters (C8-C16) and retained much of its activity in the presence of Tween-80 and Trition X-100. Lipase activity was stimulated by Mg
Sigma-Aldrich
4-Nitrophenyl butyrate, ≥98%
Sigma-Aldrich
4-Nitrophenyl dodecanoate, ≥98.0% (GC)
Sigma-Aldrich
4-Nitrophenyl octanoate, ≥90.0% (GC)
Sigma-Aldrich
4-Nitrophenyl stearate, lipase substrate
Sigma-Aldrich
4-Nitrophenyl palmitate, lipase substrate