Merck
  • Absence of equilibrium cluster phase in concentrated lysozyme solutions.
Absence of equilibrium cluster phase in concentrated lysozyme solutions.
Proceedings of the National Academy of Sciences of the United States of America (2008-03-26)
Anuj Shukla, Efstratios Mylonas, Emanuela Di Cola, Stephanie Finet, Peter Timmins, Theyencheri Narayanan, Dmitri I Svergun
ABSTRACT
In colloidal systems, the interplay between the short range attraction and long-range repulsion can lead to a low density associated state consisting of clusters of individual particles. Recently, such an equilibrium cluster phase was also reported for concentrated solutions of lysozyme at low ionic strength and close to the physiological pH. Stradner et al. [(2004) Equilibrium cluster formation in concentrated protein solutions and colloids. Nature 432:492-495] found that the position of the low-angle interference peak in small-angle x-ray and neutron scattering (SAXS and SANS) patterns from lysozyme solutions was essentially independent of the protein concentration and attributed these unexpected results to the presence of equilibrium clusters. This work prompted a series of experimental and theoretical investigations, but also revealed some inconsistencies. We have repeated these experiments following the protein preparation protocols of Stradner et al. using several batches of lysozyme and exploring a broad range of concentrations, temperature and other conditions. Our measurements were done in multiple experimental sessions at three different high-resolution SAXS and SANS instruments. The low-ionic-strength lysozyme solutions displayed a clear shift in peak positions with concentration, incompatible with the presence of the cluster phase but consistent with the system of repulsively interacting individual lysozyme molecules. Within the decoupling approximation, the experimental data can be fitted using an effective interparticle interaction potential involving short-range attraction and long-range repulsion.
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg