Merck
  • Inhibition of Cyclosporine A or rapamycin on T lymphocyte counts and the influence on the immune responses of B lymphocytes in flounder (Paralichthys olivaceus).
Inhibition of Cyclosporine A or rapamycin on T lymphocyte counts and the influence on the immune responses of B lymphocytes in flounder (Paralichthys olivaceus).
Fish & shellfish immunology (2017-05-10)
Jing Xing, Yue'e Xiao, Xiaoqian Tang, Xiuzhen Sheng, Wenbin Zhan
ABSTRACT
In acquired immunity, T lymphocytes regulate the immune responses of B lymphocytes, including the IgM
Sigma-Aldrich
p-Nitrophenyl Phosphate Liquid Substrate System, liquid
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), biotin conjugate, Type IV, lyophilized powder
Sigma-Aldrich
Hemocyanin from Megathura crenulata (keyhole limpet)