Merck
  • Photosynthetic poly-β-hydroxybutyrate accumulation in unicellular cyanobacterium Synechocystis sp. PCC 6714.
Photosynthetic poly-β-hydroxybutyrate accumulation in unicellular cyanobacterium Synechocystis sp. PCC 6714.
AMB Express (2017-07-09)
Donya Kamravamanesh, Stefan Pflügl, Winfried Nischkauer, Andreas Limbeck, Maximilian Lackner, Christoph Herwig
ABSTRACT
Poly-β-hydroxybutyrate (PHB) production from CO
Sigma-Aldrich
Crotonic acid, 98%