Merck
  • Antimetastatic effect of sulfamate carbonic anhydrase IX inhibitors in breast carcinoma xenografts.
Antimetastatic effect of sulfamate carbonic anhydrase IX inhibitors in breast carcinoma xenografts.
Journal of medicinal chemistry (2012-05-25)
Roben G Gieling, Muhammad Babur, Lupti Mamnani, Natalie Burrows, Brian A Telfer, Fabrizio Carta, Jean-Yves Winum, Andrea Scozzafava, Claudiu T Supuran, Kaye J Williams
ABSTRACT
A panel of compounds belonging to the underexposed sulfamate class of carbonic anhydrase (CA, EC 4.2.1.1) inhibitors was generated that displayed high specificity at nanomolar levels for the tumor-associated CA IX/XII isoforms. Three of the specific CA IX/XII inhibitors showed a positive response in in vitro assays for tumor cell migration and spreading. One of them, 4-(3'-(3″,5″-dimethylphenyl)ureido)phenyl sulfamate (S4), was taken forward into the orthotopic MDA-MB-231 (breast carcinoma) model in mice. Treatment with a 10 mg/kg maintenance dosage of S4 given daily on a "5 days on, 2 days off" regimen reduced metastatic tumor burden in the lung while not affecting primary tumor growth or mouse condition. CA inhibitors of the sulfamate class specifically targeting the tumor-associated isoforms are potential candidates in antimetastatic therapy.
Sigma-Aldrich
S4, ≥98% (HPLC)