Merck
  • Ubiquitin-specific peptidase 42 (USP42) functions to deubiquitylate histones and regulate transcriptional activity.
Ubiquitin-specific peptidase 42 (USP42) functions to deubiquitylate histones and regulate transcriptional activity.
The Journal of biological chemistry (2014-10-23)
Andreas K Hock, Arnaud M Vigneron, Karen H Vousden
ABSTRACT
Ubiquitin-specific peptidase 42 (USP42) is a deubiquitylating enzyme that can target p53 and contribute to the stabilization of p53 in response to stress. We now show that USP42 can also regulate transcription independently of p53. USP42 co-localized with RNA polymerase II (RNA Pol II) in nuclear foci, bound to histone H2B, and deubiquitylated H2B. Depletion of USP42 increased H2B ubiquitylation at a model promoter and decreased both basal and induced transcription from a number of promoters. These results are consistent with a role for USP42 in regulating transcription by deubiquitylating histones.
Sigma-Aldrich
KOD Hot Start Master Mix, Ready-to-use 2X mixture, containing KOD Hot Start DNA Polymerase, two monoclonal antibodies, ultrapure deoxynucleotides, and reaction buffer with MgSO4, optimized for convenient high fidelity PCR., ready-to-use solution, suitable for PCR
Sigma-Aldrich
Anti-ubiquityl-Histone H2A Antibody, clone E6C5, clone E6C5, Upstate®, from mouse
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)