Merck
  • SMAD3 Regulates Follicle-stimulating Hormone Synthesis by Pituitary Gonadotrope Cells
SMAD3 Regulates Follicle-stimulating Hormone Synthesis by Pituitary Gonadotrope Cells
The Journal of biological chemistry (2016-12-21)
Yining Li, Gauthier Schang, Ulrich Boehm, Chu-Xia Deng, Jonathan Graff, Daniel J Bernard
ABSTRACT
Pituitary follicle-stimulating hormone (FSH) is an essential regulator of fertility in females and of quantitatively normal spermatogenesis in males. Pituitary-derived activins are thought to act as major stimulators of FSH synthesis by gonadotrope cells.
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, lyophilized powder (essentially salt-free), 750-3000 units/mg solid
Sigma-Aldrich
Chorionic gonadotropin human, lyophilized powder, vial of ~2,500 IU
Sigma-Aldrich
Medium 199, With Hanks′ salts and sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, 1× crystallized, ≥97% (agarose gel electrophoresis)
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
2,2,2-Tribromoethanol, 97%