Merck
  • AlphaB-crystallin regulates remyelination after peripheral nerve injury.
AlphaB-crystallin regulates remyelination after peripheral nerve injury.
Proceedings of the National Academy of Sciences of the United States of America (2017-02-01)
Erin-Mai F Lim, Stan T Nakanishi, Vahid Hoghooghi, Shane E A Eaton, Alexandra L Palmer, Ariana Frederick, Jo A Stratton, Morgan G Stykel, Patrick J Whelan, Douglas W Zochodne, Jeffrey Biernaskie, Shalina S Ousman
ABSTRACT
AlphaB-crystallin (αBC) is a small heat shock protein that is constitutively expressed by peripheral nervous system (PNS) axons and Schwann cells. To determine what role this crystallin plays after peripheral nerve damage, we found that loss of αBC impaired remyelination, which correlated with a reduced presence of myelinating Schwann cells and increased numbers of nonmyelinating Schwann cells. The heat shock protein also seems to regulate the cross-talk between Schwann cells and axons, because expected changes in neuregulin levels and ErbB2 receptor expression after PNS injury were disrupted in the absence of αBC. Such dysregulations led to defects in conduction velocity and motor and sensory functions that could be rescued with therapeutic application of the heat shock protein in vivo. Altogether, these findings show that αBC plays an important role in regulating Wallerian degeneration and remyelination after PNS injury.
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Sigma-Aldrich
Anti-Growth Associated Protein 43 Antibody, clone 9-1E12, clone 9-1E12, Chemicon®, from mouse
Sigma-Aldrich
Anti-alpha B-crystallin Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-phosho-erbB2/Her2 (Tyr1112) Antibody, clone 19G5, clone 19G5, Upstate®, from mouse
Sigma-Aldrich
Anti-Neuregulin-1 Antibody, clone D11, clone D11, from mouse
Sigma-Aldrich
Monoclonal Anti-Neurofilament 200 (Phos. and Non-Phos.) antibody produced in mouse, clone N52, ascites fluid
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Myelin Protein P0 Antibody, from rabbit, purified by affinity chromatography