Merck
  • CDK4/6-dependent activation of DUB3 regulates cancer metastasis through SNAIL1.
CDK4/6-dependent activation of DUB3 regulates cancer metastasis through SNAIL1.
Nature communications (2017-01-10)
Tongzheng Liu, Jia Yu, Min Deng, Yujiao Yin, Haoxing Zhang, Kuntian Luo, Bo Qin, Yunhui Li, Chenming Wu, Tao Ren, Yang Han, Peng Yin, JungJin Kim, SeungBaek Lee, Jing Lin, Lizhi Zhang, Jun Zhang, Somaira Nowsheen, Liewei Wang, Judy Boughey, Matthew P Goetz, Jian Yuan, Zhenkun Lou
ABSTRACT
Tumour metastasis, the spread of cancer cells from the original tumour site followed by growth of secondary tumours at distant organs, is the primary cause of cancer-related deaths and remains poorly understood. Here we demonstrate that inhibition of CDK4/6 blocks breast tumour metastasis in the triple-negative breast cancer model, without affecting tumour growth. Mechanistically, we identify a deubiquitinase, DUB3, as a target of CDK4/6; CDK4/6-mediated activation of DUB3 is essential to deubiquitinate and stabilize SNAIL1, a key factor promoting epithelial-mesenchymal transition and breast cancer metastasis. Overall, our study establishes the CDK4/6-DUB3 axis as an important regulatory mechanism of breast cancer metastasis and provides a rationale for potential therapeutic interventions in the treatment of breast cancer metastasis.
Sigma-Aldrich
MISSION® esiRNA, targeting human CDK6
Sigma-Aldrich
Monoclonal Anti-MYC , (C-terminal) antibody produced in mouse, clone 9E10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human USP17L2