Merck
Airway Epithelial KIF3A Regulates Th2 Responses to Aeroallergens.
Journal of immunology (Baltimore, Md. : 1950) (2016-11-01)
Premkumar Vummidi Giridhar, Sheila M Bell, Anusha Sridharan, Priya Rajavelu, Joseph A Kitzmiller, Cheng-Lun Na, Matthew Kofron, Eric B Brandt, Mark Ericksen, Anjaparavanda P Naren, Changsuk Moon, Gurjit K Khurana Hershey, Jeffrey A Whitsett
ABSTRACT
KIF3A, the gene encoding kinesin family member 3A, is a susceptibility gene locus associated with asthma; however, mechanisms by which KIF3A might influence the pathogenesis of the disorder are unknown. In this study, we deleted the mouse Kif3a gene in airway epithelial cells. Both homozygous and heterozygous Kif3a gene-deleted mice were highly susceptible to aeroallergens from Aspergillus fumigatus and the house dust mite, resulting in an asthma-like pathology characterized by increased goblet cell metaplasia, airway hyperresponsiveness, and Th2-mediated inflammation. Deletion of the Kif3a gene increased the severity of pulmonary eosinophilic inflammation and expression of cytokines (Il-4, Il-13, and Il-17a) and chemokine (Ccl11) RNAs following pulmonary exposure to Aspergillus extract. Inhibition of Kif3a disrupted the structure of motile cilia and impaired mucociliary clearance, barrier function, and epithelial repair, demonstrating additional mechanisms by which deficiency of KIF3A in respiratory epithelial cells contributes to pulmonary pathology. Airway epithelial KIF3A suppresses Th2 pulmonary inflammation and airway hyperresponsiveness following aeroallergen exposure, implicating epithelial microtubular functions in the pathogenesis of Th2-mediated lung pathology.
Sigma-Aldrich
Anti-Acetylated Tubulin antibody, Mouse monoclonal, clone 6-11B-1, purified from hybridoma cell culture
Sigma-Aldrich
Albumin–fluorescein isothiocyanate conjugate, protein bovine
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human KIF3A