Merck
  • eEF-2 kinase is a critical regulator of Warburg effect through controlling PP2A-A synthesis.
eEF-2 kinase is a critical regulator of Warburg effect through controlling PP2A-A synthesis.
Oncogene (2016-05-18)
Y Cheng, X Ren, Y Yuan, Y Shan, L Li, X Chen, L Zhang, Y Takahashi, J W Yang, B Han, J Liao, Y Li, H Harvey, A Ryazanov, G P Robertson, G Wan, D Liu, A F Chen, Y Tao, J-M Yang
ABSTRACT
Cancer cells predominantly metabolize glucose by glycolysis to produce energy in order to meet their metabolic requirement, a phenomenon known as Warburg effect. Although Warburg effect is considered a peculiarity critical for survival and proliferation of cancer cells, the regulatory mechanisms behind this phenomenon remain incompletely understood. We report here that eukaryotic elongation factor-2 kinase (eEF-2K), a negative regulator of protein synthesis, has a critical role in promoting glycolysis in cancer cells. We showed that deficiency in eEF-2K significantly reduced the uptake of glucose and decreased the productions of lactate and adenosine triphosphate in tumor cells and in the Ras-transformed mouse embryonic fibroblasts. We further demonstrated that the promotive effect of eEF-2K on glycolysis resulted from the kinase-mediated restriction of synthesis of the protein phosphatase 2A-A (PP2A-A), a key factor that facilitates the ubiquitin-proteasomal degradation of c-Myc protein, as knockdown of eEF-2K expression led to a significant increase in PP2A-A protein synthesis and remarkable downregulation of c-Myc and pyruvate kinase M2 isoform, the key glycolytic enzyme transcriptionally activated by c-Myc. In addition, depletion of eEF-2K reduced the ability of the transformed cells to proliferate and enhanced the sensitivity of tumor cells to chemotherapy both in vitro and in vivo. These results, which uncover a role of the eEF-2K-mediated control of PP2A-A in tumor cell glycolysis, provide new insights into the regulation of the Warburg effect.
Sigma-Aldrich
MISSION® esiRNA, targeting human EEF2K
Sigma-Aldrich
2-Deoxy-2-[(7-nitro-2,1,3-benzoxadiazol-4-yl)amino]-D-glucose, ≥97% (HPLC)
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)