Merck
  • Simultaneous Analysis of Free and Sulfated Steroids by Liquid Chromatography/Mass Spectrometry with Selective Mass Spectrometric Scan Modes and Polarity Switching.
Simultaneous Analysis of Free and Sulfated Steroids by Liquid Chromatography/Mass Spectrometry with Selective Mass Spectrometric Scan Modes and Polarity Switching.
Analytical chemistry (2016-12-10)
Su Hyeon Lee, Nanhee Lee, Youngmin Hong, Bong Chul Chung, Man Ho Choi
ABSTRACT
Sulfated steroids can act as a latent form of active free steroids, coexisting with them in biological specimens. To evaluate the metabolic significance of free and sulfated steroid species, a simultaneous analysis of eight free steroids [cholesterol, pregnenolone, 17α-hydroxypregnenolone, progesterone, 17α-hydroxyprogesterone, dehydroepiandrosterone (DHEA), testosterone, and 17β-estradiol] and four biologically relevant sulfated steroids was developed and validated, using selected-ion and multiple-reaction monitoring modes coupled to polarity-switching liquid chromatography/mass spectrometry (LC/MS). All steroids were separated on a reversed-phase phenyl column (50 mm × 2 mm, 3 μm) at a flow rate of 0.5 mL/min. The limits of quantification ranged from 0.1 to 50 ng/mL at extraction recoveries of 94.1-105.5%, while the precision and accuracy were 2.5-9.3% and 92.4-105.9%, respectively. Quantitative results obtained for samples from obese girls showed that the serum levels of DHEA sulfate were significantly increased (P = 0.004), along with the metabolic ratio representing DHEA sulfotransferase (P < 0.02). The developed novel LC/MS method can quantitatively profile both free and sulfated steroids in a single analytical run.
Sigma-Aldrich
Dehydroepiandrosterone-2,2,3,4,4,6-d6, 97 atom % D, 98% (CP)
Sigma-Aldrich
Cholesterol-2,2,3,4,4,6-d6, 97 atom % D, 98% (CP)