Merck
  • Differential neuronal vulnerability identifies IGF-2 as a protective factor in ALS.
Differential neuronal vulnerability identifies IGF-2 as a protective factor in ALS.
Scientific reports (2016-05-18)
Ilary Allodi, Laura Comley, Susanne Nichterwitz, Monica Nizzardo, Chiara Simone, Julio Aguila Benitez, Ming Cao, Stefania Corti, Eva Hedlund
ABSTRACT
The fatal disease amyotrophic lateral sclerosis (ALS) is characterized by the loss of somatic motor neurons leading to muscle wasting and paralysis. However, motor neurons in the oculomotor nucleus, controlling eye movement, are for unknown reasons spared. We found that insulin-like growth factor 2 (IGF-2) was maintained in oculomotor neurons in ALS and thus could play a role in oculomotor resistance in this disease. We also showed that IGF-1 receptor (IGF-1R), which mediates survival pathways upon IGF binding, was highly expressed in oculomotor neurons and on extraocular muscle endplate. The addition of IGF-2 induced Akt phosphorylation, glycogen synthase kinase-3β phosphorylation and β-catenin levels while protecting ALS patient motor neurons. IGF-2 also rescued motor neurons derived from spinal muscular atrophy (SMA) patients from degeneration. Finally, AAV9::IGF-2 delivery to muscles of SOD1(G93A) ALS mice extended life-span by 10%, while preserving motor neurons and inducing motor axon regeneration. Thus, our studies demonstrate that oculomotor-specific expression can be utilized to identify candidates that protect vulnerable motor neurons from degeneration.
Sigma-Aldrich
Anti-Choline Acetyltransferase (ChAT) Antibody, serum, Chemicon®
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
Anti-Tubulin Antibody, beta III isoform, CT, clone TU-20 (Similar to TUJ1), ascites fluid, clone TU-20 (Similar to TUJ1), Chemicon®
Sigma-Aldrich
Anti-MNX1 (HB9) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Anti-phospho-Akt (pSer473) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, ascites fluid