Merck
  • Oncogenic K-ras confers SAHA resistance by up-regulating HDAC6 and c-myc expression.
Oncogenic K-ras confers SAHA resistance by up-regulating HDAC6 and c-myc expression.
Oncotarget (2016-02-06)
Qun Wang, Rong Tan, Xin Zhu, Yi Zhang, Zhiping Tan, Bing Su, Yu Li
ABSTRACT
Histone deacetylase inhibitors (HDIs) represent a new class of anticancer drugs. Suberoylanilide hydroxamic acid (SAHA), the first HDI approved for the treatment of cutaneous T cell lymphoma (CTCL), is currently being tested in clinical trials for other cancers. However, SAHA has been ineffective against solid tumors in many clinical trials. A better understanding of molecular mechanisms of SAHA resistance may provide the basis for improved patient selection and the enhancement of clinical efficacy. Here we demonstrate that oncogenic K-ras contributes to SAHA resistance by upregulating HDAC6 and c-myc expression. We find that the high levels of HDAC6 expression are associated with activated K-ras mutant in colon cancer patients. And expressions of HDAC6 and c-myc are increased in fibroblasts transformed with activated K-ras. Surprisingly, we find that activated K-ras transformed cells are more resistant to SAHA inhibition on cell growth and anchorage-independent colony formation. We show that a K-ras inhibitor sensitizes K-ras mutated lung cancer cells to SAHA induced growth inhibition. We also find that mutant K-ras induces HDAC6 expression by a MAP kinase dependent pathway. Our study suggests that combined treatment with SAHA and K-ras inhibitors may represent an effective strategy to overcome SAHA resistance.
Sigma-Aldrich
HDAC6 H216A human, recombinant, expressed in baculovirus infected Sf9 cells, ≥75% (SDS-PAGE)
Sigma-Aldrich
HDAC6 H611A human, recombinant, expressed in baculovirus infected Sf9 cells, ≥79% (SDS-PAGE)