Merck
  • Noncanonical activation of Akt/protein kinase B in {beta}-cells by the incretin hormone glucose-dependent insulinotropic polypeptide.
Noncanonical activation of Akt/protein kinase B in {beta}-cells by the incretin hormone glucose-dependent insulinotropic polypeptide.
The Journal of biological chemistry (2009-02-24)
Scott B Widenmaier, Arthur V Sampaio, T Michael Underhill, Christopher H S McIntosh
ABSTRACT
Therapeutics based on the actions of the incretin hormones, glucagon-like peptide-1 (GLP-1) and glucose-dependent insulinotropic polypeptide (GIP), have recently been introduced for the treatment of type 2 diabetes mellitus. The serine/threonine kinase Akt is a major mediator of incretin action on the pancreatic islet, increasing beta-cell mass and function and promoting beta-cell survival. The mechanisms underlying incretin activation of Akt are thought to involve an essential phosphoinositide 3-kinase-mediated phosphorylation of threonine 308, similar to the prototypical Akt activator, insulin-like growth factor-I (IGF-I). In this study, using activity assays on immunoprecipitated Akt, we discovered that GIP and GLP-1 were capable of stimulating Akt in the INS-1 beta-cell line and isolated mouse islets via a mechanism that did not require phosphoinositide 3-kinase or phosphorylation of Thr(308) and Ser(473), and this pathway involved the production of cAMP. Furthermore, we found that GIP stimulated anti-apoptotic signaling via this alternate mode of Akt activation. We conclude that incretins can activate Akt via a novel noncanonical mechanism that may provide an alternative therapeutic target for the treatment of type 2 diabetes mellitus and have broader implications for Akt physiology in human health and disease.
Sigma-Aldrich
Monoclonal Anti-MDM2 antibody produced in mouse, clone SMP14, ascites fluid
Pricing and availability is not currently available.