Merck
  • A 3'UTR-associated RNA, FLJ11812 maintains stemness of human embryonic stem cells by targeting miR-4459.
A 3'UTR-associated RNA, FLJ11812 maintains stemness of human embryonic stem cells by targeting miR-4459.
Stem cells and development (2014-12-02)
Wei Lu, Lei Han, Le Su, Jing Zhao, Yun Zhang, ShangLi Zhang, BaoXiang Zhao, JunYing Miao
ABSTRACT
The 3' untranslated region (UTR)-associated RNAs (uaRNAs) have important roles in various biological processes, especially in development. However, since they overlap with protein-coding mRNAs, uaRNAs are difficult to study by RNA interference techniques. We recently identified a chemical molecule, 3-benzyl-5-((2-nitrophenoxy) methyl)-dihydrofuran-2(3H)-one (3BDO), that could efficiently induce human embryonic stem cells (hESCs) differentiation, and meanwhile selectively and efficiently downregulate the uaRNA FLJ11812. By acting as a competing endogenous RNA, downregulated FLJ11812 by 3BDO further increased miR-4459 level in hESCs. miR-4459 could decrease the expression of its targets, CDC20B and ATG13, and thus altered stemness via cell cycle and autophagy. Our results revealed that FLJ11812 played a key role in maintenance of stemness of hESCs for the first time. The findings provide new clues and a powerful tool for investigating the action mechanism of FLJ11812 in early development.
Sigma-Aldrich
Anti-ATG13 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody