Merck
  • Antidepressants inhibit DNA methyltransferase 1 through reducing G9a levels.
Antidepressants inhibit DNA methyltransferase 1 through reducing G9a levels.
The Biochemical journal (2012-08-14)
Nicole Zimmermann, Jürgen Zschocke, Tatjana Perisic, Shuang Yu, Florian Holsboer, Theo Rein
ABSTRACT
The discovery of epigenetic processes as possible pivotal regulatory mechanisms in psychiatric diseases raised the question of how psychoactive drugs may impact the epigenetic machinery. In the present study we set out to explore the specificity and the mode of action of the reported inhibitory effect of the TCA (tricyclic antidepressant) amitriptyline on DNMT (DNA methyltransferase) activity in primary astrocytes from the rat cortex. We found that the impact on DNMT was shared by another TCA, imipramine, and by paroxetine, but not by venlafaxine or the mood stabilizers carbamazepine and valproic acid. DNMT activity in subventricular neural stem cells was refractory to the action of ADs (antidepressants). Among the established DNMTs, ADs primarily targeted DNMT1. The reduction of enzymatic DNMT1 activity was neither due to reduced DNMT1 expression nor due to direct drug interference. We tested putative DNMT1-inhibitory mechanisms and discovered that a known stimulator of DNMT1, the histone methyltransferase G9a, exhibited decreased protein levels and interactions with DNMT1 upon AD exposure. Adding recombinant G9a completely reversed the AD repressive effect on DNMT1 function. In conclusion, the present study presents a model where distinct ADs affect DNMT1 activity via G9a with important repercussions for possible novel treatment regimes.
Sigma-Aldrich
Gö 6983, A potent, cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase C (PKC) that inhibits several PKC isozymes.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-ARD1A antibody produced in rabbit, affinity isolated antibody