Merck
  • Histone deacetylase inhibitors down-regulate bcl-2 expression and induce apoptosis in t(14;18) lymphomas.
Histone deacetylase inhibitors down-regulate bcl-2 expression and induce apoptosis in t(14;18) lymphomas.
Molecular and cellular biology (2005-02-17)
Hong Duan, Caroline A Heckman, Linda M Boxer
ABSTRACT
Histone deacetylase (HDAC) inhibitors are promising antitumor agents, but they have not been extensively explored in B-cell lymphomas. Many of these lymphomas have the t(14;18) translocation, which results in increased bcl-2 expression and resistance to apoptosis. In this study, we examined the effects of two structurally different HDAC inhibitors, trichostatin A (TSA) and sodium butyrate (NaB), on the cell cycle, apoptosis, and bcl-2 expression in t(14;18) lymphoma cells. We found that in addition to potent cell cycle arrest, TSA and NaB also dramatically induced apoptosis and down-regulated bcl-2 expression, and overexpression of bcl-2 inhibited TSA-induced apoptosis. The repression of bcl-2 by TSA occurred at the transcriptional level. Western blot analysis and quantitative chromatin immunoprecipitation (ChIP) assay showed that even though HDAC inhibitors increased overall acetylation of histones, localized histone H3 deacetylation occurred at both bcl-2 promoters. TSA treatment increased the acetylation of the transcription factors Sp1 and C/EBPalpha and decreased their binding as well as the binding of CBP and HDAC2 to the bcl-2 promoters. Mutation of Sp1 and C/EBPalpha binding sites reduced the TSA-induced repression of bcl-2 promoter activity. This study provides a mechanistic rationale for the use of HDAC inhibitors in the treatment of human t(14;18) lymphomas.
Sigma-Aldrich
Anti-acetyl-Histone H4 Antibody, serum, Upstate®
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-acetyl-Lysine Antibody, clone 4G12, clone 4G12, Upstate®, from mouse
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit