Merck
  • Effects of lead on the morphology and structure of the nucleolus in the root tip meristematic cells of Allium cepa L.
Effects of lead on the morphology and structure of the nucleolus in the root tip meristematic cells of Allium cepa L.
International journal of molecular sciences (2014-08-05)
Ze Jiang, Huaning Zhang, Rong Qin, Jinhua Zou, Junran Wang, Qiuyue Shi, Wusheng Jiang, Donghua Liu
ABSTRACT
To study the toxic mechanisms of lead (Pb) in plants, the effects of Pb on the morphology and structure of the nucleolus in root tip meristematic cells of Allium cepa var. agrogarum L. were investigated. Fluorescence labeling, silver-stained indirect immunofluorescent microscopy and western blotting were used. Fluorescence labeling showed that Pb ions were localized in the meristematic cells and the uptake and accumulation of Pb increased with treatment time. At low concentrations of Pb (1-10 μM) there were persistent nucleoli in some cells during mitosis, and at high concentration (100 μM) many of the nucleolar organizing regions were localized on sticky chromosomes in metaphase and anaphase cells. Pb induced the release of particles containing argyrophilic proteins to be released from the nucleus into the cytoplasm. These proteins contained nucleophosmin and nucleolin. Pb also caused the extrusion of fibrillarin from the nucleus into the cytoplasm. Western blotting demonstrated the increased expression of these three major nucleolar proteins under Pb stress.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)−FITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution