Merck
  • A Phase I study of an intravesically administered immunotoxin targeting EpCAM for the treatment of nonmuscle-invasive bladder cancer in BCGrefractory and BCG-intolerant patients.
A Phase I study of an intravesically administered immunotoxin targeting EpCAM for the treatment of nonmuscle-invasive bladder cancer in BCGrefractory and BCG-intolerant patients.
Drug design, development and therapy (2010-12-15)
Mark Kowalski, Joycelyn Entwistle, Jeannick Cizeau, Demi Niforos, Shauna Loewen, Wendy Chapman, Glen C MacDonald
ABSTRACT
A Phase I dose-escalation study was performed to determine the maximum tolerated dose (MTD) of the immunotoxin VB4-845 in patients with nonmuscle-invasive bladder cancer (NMIBC) refractory to or intolerant of bacillus Calmette-Guerin (BCG). Secondary objectives included evaluation of the safety, tolerability, pharmacokinetics, immunogenicity, and efficacy of VB4-845. Sixty-four patients with Grade 2 or 3, stage Ta or T1 transitional cell carcinoma or in situ carcinoma, either refractory to or intolerant of BCG therapy, were enrolled. Treatment was administered in ascending dose cohorts ranging from 0.1 to 30.16 mg. After receiving weekly instillations of VB4-845 to the bladder via catheter for 6 consecutive weeks, patients were followed for 4-6 weeks post-therapy and assessed at week 12. An MTD was not determined, as a dose-limiting toxicity was not identified over the dose range tested. VB4-845 therapy was safe and well tolerated with most adverse events reported as mild; as a result, no patients were removed from the study in response to toxicity. By the end of the study, the majority of patients had developed antibodies to the exotoxin portion of VB4-845. A complete response was achieved in 39% of patients at the 12-week time point. VB4-845 dosed on a weekly basis for 6 weeks was very well tolerated at all dose levels. Although an MTD was not determined at the doses administered, VB4-845 showed evidence of an antitumor effect that warrants further clinical investigation for the treatment of NMIBC in this patient population.
Sigma-Aldrich
Anti-Pseudomonas Exotoxin A antibody produced in rabbit, whole antiserum