Merck
  • Excess centrosomes perturb dynamic endothelial cell repolarization during blood vessel formation.
Excess centrosomes perturb dynamic endothelial cell repolarization during blood vessel formation.
Molecular biology of the cell (2016-04-22)
Erich J Kushner, Luke S Ferro, Zhixian Yu, Victoria L Bautch
ABSTRACT
Blood vessel formation requires dynamic movements of endothelial cells (ECs) within sprouts. The cytoskeleton regulates migratory polarity, and centrosomes organize the microtubule cytoskeleton. However, it is not well understood how excess centrosomes, commonly found in tumor stromal cells, affect microtubule dynamics and interphase cell polarity. Here we find that ECs dynamically repolarize during sprouting angiogenesis, and excess centrosomes block repolarization and reduce migration and sprouting. ECs with excess centrosomes initially had more centrosome-derived microtubules but, paradoxically, fewer steady-state microtubules. ECs with excess centrosomes had elevated Rac1 activity, and repolarization was rescued by blockade of Rac1 or actomyosin blockers, consistent with Rac1 activity promoting cortical retrograde actin flow and actomyosin contractility, which precludes cortical microtubule engagement necessary for dynamic repolarization. Thus normal centrosome numbers are required for dynamic repolarization and migration of sprouting ECs that contribute to blood vessel formation.
Sigma-Aldrich
Anti-GM130 (C-terminal) antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Puromycin dihydrochloride, Ready Made Solution, from Streptomyces alboniger, 10 mg/mL in H2O, suitable for cell culture
Sigma-Aldrich
Anti-α-Tubulin Antibody, clone DM1A, Alexa Fluor 488 conjugate, clone DM1A, Upstate®, from mouse
Sigma-Aldrich
Anti-α-Tubulin Antibody, Alexa Fluor 555 conjugate, clone DM1A, from mouse, ALEXA FLUOR 555