Merck
  • Multiresidue analysis of cephalosporin antibiotics in bovine milk based on molecularly imprinted polymer extraction followed by liquid chromatography-tandem mass spectrometry.
Multiresidue analysis of cephalosporin antibiotics in bovine milk based on molecularly imprinted polymer extraction followed by liquid chromatography-tandem mass spectrometry.
Journal of chromatography. A (2016-11-07)
A N Baeza, J L Urraca, R Chamorro, G Orellana, M Castellari, M C Moreno-Bondi
ABSTRACT
This work reports the preparation of molecularly imprinted polymers (MIPs) selective to cephalosporin (CF) antibiotics, and their application as molecularly imprinted solid-phase extraction (MISPE) sorbents for the determination of these antimicrobials in milk samples. Several functional monomers and cross-linkers have been screened to select the best combination that provides high selectivity for the simultaneous multiresidue extraction of cefthiofur (THIO), cefazolin (AZO), cefquinome (QUI), cephapirin (API), cephalexin (ALE) and cephalonium (ALO) from the samples. The novel MIPs were prepared by a non-covalent imprinting approach in the form of spherical microparticles using the synthetic surrogate molecule sodium 7-(2-biphenylylcarboxamido)-3-methyl-3-cepheme-4-carboxylate, N-3,5-bis(trifluoromethyl)phenyl-N'-4-vinylphenyl urea (VPU) as functional monomer, and divinylbenzene (DVB) as crosslinking agent in a 1:2:20 molar ratio. The optimized MISPE method allowed the extraction of the target antimicrobials from raw cow milk samples using a selective washing with 5mL methanol/2-[4-(2-hydroxyethyl)-1-piperazinyl]ethanesulfonic acid (HEPES) buffer (0.1M, pH 7.5) (2:98, v/v) to remove the non-specifically retained compounds, followed by elution with 1mL of trifluoroacetic acid (TFA) in methanol (0.1:99.9, v/v). The extracts have been analysed by UHPLC-MS/MS and the analytical method has been validated according to EU guideline 2002/657/EC. The limits of quantification (S/N=10) were in the 1.7-12.5μgkg
Product Description
Supelco
Cefquinome sulfate, VETRANAL®, analytical standard
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole
Sigma-Aldrich
2-(Diethylamino)ethyl methacrylate, contains 1500 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Trimethylolpropane trimethacrylate, contains 250 ppm monomethyl ether hydroquinone as inhibitor, technical grade