Merck
  • The Robo4 cytoplasmic domain is dispensable for vascular permeability and neovascularization.
The Robo4 cytoplasmic domain is dispensable for vascular permeability and neovascularization.
Nature communications (2016-11-25)
Feng Zhang, Claudia Prahst, Thomas Mathivet, Laurence Pibouin-Fragner, Jiasheng Zhang, Gael Genet, Raymond Tong, Alexandre Dubrac, Anne Eichmann
ABSTRACT
Vascular permeability and neovascularization are implicated in many diseases including retinopathies and diabetic wound healing. Robo4 is an endothelial-specific transmembrane receptor that stabilizes the vasculature, as shown in Robo4
Sigma-Aldrich
MISSION® esiRNA, targeting human ROBO4
Sigma-Aldrich
MISSION® esiRNA, targeting human UNC5B
Sigma-Aldrich
Fibrinogen from bovine plasma, Type I-S, 65-85% protein (≥75% of protein is clottable)
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human ROBO2
Sigma-Aldrich
MISSION® esiRNA, targeting human ROBO1