Merck
  • NMDAR inhibition-independent antidepressant actions of ketamine metabolites.
NMDAR inhibition-independent antidepressant actions of ketamine metabolites.
Nature (2016-05-05)
Panos Zanos, Ruin Moaddel, Patrick J Morris, Polymnia Georgiou, Jonathan Fischell, Greg I Elmer, Manickavasagom Alkondon, Peixiong Yuan, Heather J Pribut, Nagendra S Singh, Katina S S Dossou, Yuhong Fang, Xi-Ping Huang, Cheryl L Mayo, Irving W Wainer, Edson X Albuquerque, Scott M Thompson, Craig J Thomas, Carlos A Zarate, Todd D Gould
ABSTRACT
Major depressive disorder affects around 16 per cent of the world population at some point in their lives. Despite the availability of numerous monoaminergic-based antidepressants, most patients require several weeks, if not months, to respond to these treatments, and many patients never attain sustained remission of their symptoms. The non-competitive, glutamatergic NMDAR (N-methyl-d-aspartate receptor) antagonist (R,S)-ketamine exerts rapid and sustained antidepressant effects after a single dose in patients with depression, but its use is associated with undesirable side effects. Here we show that the metabolism of (R,S)-ketamine to (2S,6S;2R,6R)-hydroxynorketamine (HNK) is essential for its antidepressant effects, and that the (2R,6R)-HNK enantiomer exerts behavioural, electroencephalographic, electrophysiological and cellular antidepressant-related actions in mice. These antidepressant actions are independent of NMDAR inhibition but involve early and sustained activation of AMPARs (α-amino-3-hydroxy-5-methyl-4-isoxazole propionic acid receptors). We also establish that (2R,6R)-HNK lacks ketamine-related side effects. Our data implicate a novel mechanism underlying the antidepressant properties of (R,S)-ketamine and have relevance for the development of next-generation, rapid-acting antidepressants.
Sigma-Aldrich
(2R,6R)-Hydroxynorketamine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
(2S,6S)-Hydroxynorketamine hydrochloride, ≥98% (HPLC)