- Synthesis and structure-activity relationships of N3-pyridylpyrazinones as corticotropin-releasing factor-1 (CRF1) receptor antagonists.
Synthesis and structure-activity relationships of N3-pyridylpyrazinones as corticotropin-releasing factor-1 (CRF1) receptor antagonists.
Bioorganic & medicinal chemistry letters (2010-02-24)
Richard A Hartz, Vijay T Ahuja, William D Schmitz, Thaddeus F Molski, Gail K Mattson, Nicholas J Lodge, Joanne J Bronson, John E Macor
PMID20176478
ABSTRACT
A series of N(3)-pyridylpyrazinones was investigated as corticotropin-releasing factor-1 receptor antagonists. It was observed that the binding affinity of analogues containing a pyridyl group was influenced not only by the substitution pattern on the pyridyl group, but also by the pK(a) of the pyridyl nitrogen. Analogues containing a novel 6-(difluoromethoxy)-2,5-dimethylpyridin-3-amine group were among the most potent N(3)-pyridylpyrazinones synthesized. The synthesis and SAR of N(3)-pyridylpyrazinones is described herein.